DNA fingerprinting kit
Leverandør: G-Biosciences
BE-104
BE-104EA 5250 NOK
BE-104
DNA fingerprinting kit
Lærings materiell Læremateriale for kriminologi Fingerprint Analysis Learning Activities
This DNA fingerprinting kit allows students to carry out their own criminal investigation by comparing DNA samples collected from suspects to DNA collected at a pseudo-crime scene.
  • Introduces DNA fingerprinting technique
  • Teaches DNA fragmentation by specific enzymes
  • Introduction to agarose electrophoresis
Students will use specialised molecules to cut the DNA into fragments, which can then be visualised by a process known as electrophoresis. The resulting pattern, or fingerprint, can be compared and the guilty suspect identified.