Experimentation kits, nucleic acid quantification
Leverandør: G-Biosciences
BE-318
BE-318EA 7325 NOK
BE-318
Experimentation kits, nucleic acid quantification
Lærings materiell Læremateriale for biologi Biochemistry Learning Activities
The kit utilises the principle of diffusion of nucleic acids on a nylon membrane to determine their concentration. No spectrophotometers required.
  • No expensive equipment required
  • Rapidly quantitate DNA and RNA samples
  • Enough reagents for over 50 assays
Also required: DNA samples.
Nucleic acid concentration can be measured by measuring the diameter of diffused nucleic acid spots or comparing the colour density of the spots with a set of known standards. Accurate and fast DNA and RNA concentrations can be determined with as little as 1 μl of DNAMore Product Information