Experimentation kits, physical properties of proteins
Leverandør: G-Biosciences
BE-401
BE-401EA 4750 NOK
BE-401
Experimentation kits, physical properties of proteins
Lærings materiell Læremateriale for biologi Biochemistry Learning Activities
The physical properties of proteins kit is a lab activity that enables students to investigate the physical properties of several different proteins. Students will learn about protein solubility and how it is affected by various parameters; including temperature, pH, salt and dielectric constant. They will understand about protein precipitation due to pH, high salt and in the presence of organic solvents and about protein denaturation as a result of high temperature. In addition, the kit will demonstrate how non-protein agents, such as detergents drastically alter the physical properties ofMore Product Information