Experimentation kits: biodiversity study and biomass analysis
Leverandør: G-Biosciences
BE-403
BE-403EA 4350 NOK
BE-403
Experimentation kits: biodiversity study and biomass analysis
Lærings materiell Læremateriale for biologi Biochemistry Learning Activities
Biodiversity lab activity is designed for determination of protein contents and biomass in diverse biological samples to study how biomass is related to biodiversity in nature. Students collect and catalogue plant leaf samples from a diverse group of locally available plants. This lab activity involves determination of natural weight of each plant sample, grinding a predetermined amount of each sample, and the subsequent extraction of proteins from the samples. Students then learn to determine protein contents of each plant sample and attempt to relate the protein content with biomass.More Product Information