Experimentation kits, protein fingerprinting
Leverandør: G-Biosciences
BE-408
BE-408EA 4900 NOK
BE-408
Experimentation kits, protein fingerprinting
Lærings materiell Læremateriale for biologi Biochemistry Learning Activities
In this lab activity, students learn to perform simple protein isolation procedures to isolate the protein fingerprint from various fresh tissues. They will compare the protein fingerprints of 4 different tissues to understand that the function of a particular organ is due to the proteins that are localised to the specific organ. Also included in this kit are four dried protein samples (from mouse liver, brain, heart and lungs) to compare as a control, if fresh tissues cannot be obtained.
  • Manipulation of protein samples
  • Protein molecules as the building