Experimentation kits, dialysis
Leverandør: G-Biosciences
BE-603
BE-603EA 5100 NOK
BE-603
Experimentation kits, dialysis
Lærings materiell Læremateriale for kjemi Chromatography Learning Activities
Dialysis is a routinely used technique in research laboratories to 'change' the solution a biomolecule is dissolved in. Often the buffers used to isolate biomolecules, such as proteins, are not compatible with downstream applications due to high concentrations of reagents, such as salts. A sample is placed in a bag with a semi-permeable membrane and then placed in the new solution or water. Small molecules, such as salt, can pass through the membrane, moving from an area of high concentration to low concentration. The larger molecules, such as proteins, are retained in the bag. MicrodialysisMore Product Information