GenFind V3 Reagent Kits
Leverandør: Beckman Coulter
C34880
BECKC34880EA 17000 NOK
BECKC34880
GenFind V3 Reagent Kits
Nukleinsyre reagenser Nukleinsyrerensingsreagenser og -kit
This prep system kit isolates and purifies genomic DNA (gDNA) from whole blood. The kit is uses Solid Phase Reversible Immobilisation (SPRI) paramagnetic bead-based technology to effectively produce a high recovery of high quality DNA for downstream applications such as PCR.
  • Automated available
Application: Genomic DNA extraction from whole blood.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Applikasjon
Target molecule
Refine target molecule
Sample type Blood
Method / Format