Microtitre Plates, FlowerPlate, pH/DO 2 (LG1/RF)

Leverandør: Beckman Coulter
M2P-MTP-48BOH2
BECKM2P-MTP-48BOH2EA 75275 NOK
BECKM2P-MTP-48BOH2
Microtitre Plates, FlowerPlate, pH/DO 2 (LG1/RF)
Mikroplater HTS-mikroplater
The FlowerPlate microtiter plate is a 48-well MTP in standard ANSI/SLAS (SBS) format. Its flower-shaped wells resemble baffles in shake flasks and facilitate high gas transfer into the liquid phase.

This FlowerPlate MTP has two optical sensors (optodes) per cultivation well, i.e. the LG1 optode for measuring the pH value and the RF optode for measuring the DO.

The following bioprocess parameters can be read out online by the BioLector microbioreactor:
• Biomass
• pH
• Dissolved oxygen (DO)
• Fluorescence

This MTP is compatible with: BioLector II, BioLector Pro and BioLector XT microbioreactors.
Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR