Stand for Imhoff sedimentation cones
Leverandør: BEHR
B00179003
BEHRB00179003EA 2400 NOK
BEHRB00179003
Stand for Imhoff sedimentation cones
Stativer
Rack for Sedimention vessels, made of solid PVC grey, for two Imhoff cones with stopcock.