Extraction unit for determination of hydrocarbon
Leverandør: BEHR
B00217637
BEHRB00217637EA 5300 NOK
BEHRB00217637
Extraction unit for determination of hydrocarbon
Ekstraktorer Væske-væske-uttrekkere
Behrotest® extraction unit EX1000 is used for the determination of the hydrocarbon.
  • Individual functional parts are easy to clean
  • Easy to assemble
The funnel separator facilitates the separation of the organic phase. The individual functional parts of the separator can be easily separated. The user can thus transfer the organic phase quickly and simply in one step to the clean-up column.
Sertifiseringer: The PFL sampling bottle is ideal for direct extraction in terms of ISO 9377-2.