behrotest® Clean-up station, CUS 2
Leverandør: BEHR
B00217638
BEHRB00217638EA 8275 NOK
BEHRB00217638
behrotest® Clean-up station, CUS 2
Destruksjonssystemer Arbeidsstasjoner for Mineraliseirng
The complete sample clean up station includes dropping funnel, clean-up column with glass frit, reducing flasks and stand.
The clean up procedure can be done for two samples simultaneously.
Sertifiseringer: Corresponds exactly to the provisions of ISO 9377-2.