behrotest® nitrogen station, KOSTA 2
Leverandør: BEHR
B00217639
BEHRB00217639EA 7150 NOK
BEHRB00217639
behrotest® nitrogen station, KOSTA 2
Destruksjonssystemer Arbeidsstasjoner for Mineraliseirng
In the behrotest® nitrogen station, solvent is evaporated under ambient temperature conditions by using a mild nitrogen flow. A distributor leads the nitrogen flow through two individually adjustable tubes into the Kuderna-Danish flasks. Thus the two samples reduce simulataneously.
The organic phase can be reduced down to less than 1 ml under a mild nitrogen flow.
Sertifiseringer: ISO 9377-2