Compact system for 500 ml extraction KEX 500 / KEX 500 F, behrotest®
Leverandør: BEHR
B00217739
BEHRB00217739EA 0 NOK
BEHRB00217739
Compact system for 500 ml extraction KEX 500 / KEX 500 F, behrotest®
Anaerobe systemer
behrotest® serial extraction devices are the economical and user-friendly solution for classic Soxhlet and fat extraction.
With behr hydrolysis units, acid digestion is possible before extraction (determination of total fat content according to Weibull and Stoldt).
  • Complete with reaction vessels, extraction spacers and coolers
  • The heating mantle in use is continuously variable and has an integrated magnetic stirrer.
  • Practical stand for holding condensers securely between extractions
  • Holders for secure fixation of extraction adapters after removal of the