behrosog 3 neutraliser
Leverandør: BEHR
B00217925
BEHRB00217925EA 54625 NOK
BEHRB00217925
behrosog 3 neutraliser
Destruksjonssystemer Arbeidsstasjoner for Mineraliseirng
The two-level behrosog process extraction system (neutraliser) – pre-separator plus safety level – keeps all acid vapours entirely away from the environment.
  • Efficient operating costs
  • Infinitely regulated extraction through-flow
  • Both cleaning levels (condensation and neutralisation level) prevent the emission of toxic substances into the environment
Compact process extraction system for extracting and neutralising aggressive acid vapours, especially from the Kjeldahl digestion for nitrogen determination. An upstream two-level pre-separator washes theMore Product Information