Workstations for heavy metal sample analyses, behrotest® SMA-ARM
Leverandør: BEHR
B00218413
BEHRB00218413EA 0 NOK
BEHRB00218413
Workstations for heavy metal sample analyses, behrotest® SMA-ARM
Destruksjonssystemer Arbeidsstasjoner for Mineraliseirng
The behrotest® SMA-ARM workstation, for simultaneous aqua regia digestion of up to six samples for heavy metals analysis, are supplied with a programmable microprocessor-controlled time/temperature controller as well as a fume manifold for drawing off the acid vapours. This results in greater convenience and functionality in routine laboratory operations.
The details of the components of the workstation are described in the table below: