S serial extraction units, R 104 S/R 104 S-SK, behrotest®
Leverandør: BEHR
B00218425
BEHRB00218425EA 50275 NOK
BEHRB00218425
S serial extraction units, R 104 S/R 104 S-SK, behrotest®
Ekstraktorer Soxhlet- og Soxhlet-typeuttrekkere
Behr equipment for Soxhlet extraction comply with the most various requirements of daily laboratory routines.
  • Practical holders for condenser and extraction adapter for secure placement between extractions
  • Extractor sizes from 30 ml to 1000 ml
  • Condenser with screw connections
Complete compact extraction system with basic rack, heating device, holder, hoses and glassware (reaction flasks, extractor, Dimroth condenser for extraction). Infinitely variable heating regulation After the extraction cycle, the compact system removes the solvent, by means of aMore Product Information