Digestion system for hydroxyproline, behrotest®
Leverandør: BEHR
B00218426
BEHRB00218426EA 0 NOK
BEHRB00218426
Digestion system for hydroxyproline, behrotest®
Destruksjonssystemer Arbeidsstasjoner for Mineraliseirng
This six sample digestion apparatus with energy control is suitable for determination of hydroxyproline content.