H-B DURAC® Plus Liquid-in-Glass Laboratory Thermometer
Leverandør: Bel-Art Products, a Part of SP
B60700-1500
BELAB60700-1500EA 0 NOK
BELAB60700-1500
H-B DURAC® Plus Liquid-in-Glass Laboratory Thermometer
Termometere Analogue Thermometers
Durable, partial and total immersion thermometers. Mercury free, these thermometers feature blue organic liquid fill against lead-free white back glass.
  • Traceable to NIST, individually serialised
  • Ideal for use in chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical/medical industry, dairies, breweries and distilleries
  • Include multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability
  • Tested and inspected by our USA quality team