Maximum registering, autoclave bi-metal thermometer, H-B DURAC®, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
B613150000
BELAB613150000EA 1925 NOK
BELAB613150000
Maximum registering, autoclave bi-metal thermometer, H-B DURAC®, SP Bel-Art
Termometere Analogue Thermometers
This durable bi-metal thermometer has a maximum registering pointer (red) that is moved up by the temperature measuring pointer (black) to indicate the highest temperature achieved, and remains there until manually reset.
  • Threaded connection (¼" NPT) screws directly into in-line processes, can be used to mount thermometer on included stand
  • Waterproof and pressurized, also has a removable protective silicone sleeve to prevent moisture from making contact with the dial
  • Constructed from durable 304 stainless steel with a glass lens
  • Withstands pressure, high