Dispenser, Measure-Matic™, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
F116540000
BELAF116540000EA 820 NOK
BELAF116540000
Dispenser, Measure-Matic™, SP Bel-Art
Flasketoppdispensere
Bottles, LDPE, translucent, for quick dispensing, with polypropylene measuring cup. Measuring cup slides onto a polypropylene delivery tube. The height of the tube determines the volume of liquid dispensed into the measuring cup when the bottle is squeezed.
  • Repetitive dispensing of the same volume of liquid
  • When the bottle pressure is released excess flluid returns into the bottle for spill free pouring
  • Good chemical resistance