Wash bottles, wide mouth, with coloured caps, labeled, Safety-Vented™
Leverandør: Bel-Art Products, a Part of SP
F118320019
BELAF118320019EA 1400 NOK
BELAF118320019
Wash bottles, wide mouth, with coloured caps, labeled, Safety-Vented™
Flasker Spruteflasker
LDPE, with PP spray cap (vented).
Bottles feature a 53 mm wide mouth for easy refilling. They are pre-printed with the chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment. Leak-proof, these translucent LDPE bottles have color-coded PP closures to aid in quick and easy identification.
A unique valve prevents pressure build-up inside these Right-to-Know safety-labeled wash bottles, eliminating potentially dangerous drips. This valveMore Product Information