Fluorescent ruler, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
F133550001
BELAF133550001EA 605 NOK
BELAF133550001
Fluorescent ruler, SP Bel-Art
Linjaler
Fluorescent ruler can be used in observing and photographing ethidium bromide stained nucleic acid agarose and acrylamide gels.
  • Ruler fluorescences visibly upon illumination with ultraviolet light
  • Measure of nucleic acid mobility or quantify the size of nucleic acid fragments
  • Measurement allows for comparison of gels run at different times under the same conditions