Markers, bel-pen, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
F133740000
BELAF133740000EA 695 NOK
BELAF133740000
Markers, bel-pen, SP Bel-Art
Penner Permanent Markers
These felt tip marking pens are perfect for labeling all types of glassware and plastics. The permanent and quick drying ink remains waterproof and smudge proof.
  • Will withstand autoclaving when dry
  • Pen with handy pocket clip is 133 mm long