Fritware immersion filters, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
F136700000
BELAF136700000EA 1525 NOK
BELAF136700000
Fritware immersion filters, SP Bel-Art
Filter Filterrør
Immersion filters for filtration involving recovery of solids without having to transfer material to funnels.
  • High density PE plastic coarse porosity disc and body
  • Use directly or precoat for fine filtration
  • Has a serrated tubing connector