Sponge, Cleanware™, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
F170850000
BELAF170850000EA 690 NOK
BELAF170850000
Sponge, Cleanware™, SP Bel-Art
Papirservietter Svamper
This steel wool sponge is designed to scrub glassware and stainless steel products sparkling clean with or without detergents.
  • No sharp edges to damage surfaces or irritate hands
  • Pliant sponges are flexible enough to thoroughly clean hard to reach areas
  • Measures roughly 6,4×10 cm
  • Grade 430 steel wool