Rack for cuvettes, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
F185150000
BELAF185150000EA 930 NOK
BELAF185150000
Rack for cuvettes, SP Bel-Art
Stativer for rør Kyvettestativer
Made of white PP. Rack holds 10 mm light path spectrophotometer cuvettes in two tiers with numbered wells.
  • Autoclavable
  • Very good chemical resiatnce
  • Easy to clean
The wells hold standard cuvettes with a 12,5 mm cross-section. A sloped top makes the second tier more accessible.