Racks for NMR sample tubes, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
F188540005
BELAF188540005EA 1700 NOK
BELAF188540005
Racks for NMR sample tubes, SP Bel-Art
Stativer for rør Tube Racks
These PP racks are made specifically for organising and protecting thin-walled nuclear magnetic resonance tubes.
  • Hemispherical wells in lower deck cradle tubes
  • End plate handles make transport easy
  • Racks stack securely to conserve bench space
  • Chemical resistant and autoclavable