Floating racks for cyrotubes, Bel-Art ProCulture, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
F188760020
BELAF188760020EA 855 NOK
BELAF188760020
Floating racks for cyrotubes, Bel-Art ProCulture, SP Bel-Art
Stativer for rør Tube Racks
This 20 place polypropylene round floating rack fits most cryotubes and holds them tightly in a circular pattern formed by 12 outer and 8 inner openings.
  • Floats in water; can also be used for quickly thawing frozen cells in a 37 ºC water bath
  • Easily remove tubes by holding the center handle and pushing the rack down against the bench until the legs touch
  • Dimensions: 6,4 mm (¹/₄") thick; 95 mm (3³/₄") in Ø; 19 mm (³/₄") long legs
  • Autoclavable
Excellent for holding cryotubes during work or incubation.