SP Bel-Art® dippers
Leverandør: Bel-Art Products, a Part of SP
F367840016 F367850016 F367860016
BELAF367840016EA 1625 NOK
BELAF367840016 BELAF367850016 BELAF367860016
SP Bel-Art® dippers
Prøvetakere Prøveøser
The cups have threaded fittings that screw onto the handle.
  • Two puring spouts for left- or right-handed people
  • Handles are strong honeycomb tubes without inserts and ripped for a sure grip
  • End of the handle with a hook for hanging storage
Suitable for convenient sampling from tanks, vats, streams, ponds, lakes.