Scraper, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
F368200001
BELAF368200001EA 1050 NOK
BELAF368200001
Scraper, SP Bel-Art
Skrapere
PP, white.
  • Rigid, chisel-edged, triangular blade is tapered and fans out to a width of 11,1 cm (4⅜")
  • Handle is shaped for a comfortable and secure grip
  • Steam autoclavable at 121 °C
  • Complies with FDA requirements for direct contact with foods, drugs and cosmetics