SP Bel-Art® sampling syringe
Leverandør: Bel-Art Products, a Part of SP
F378790000
BELAF378790000EA 855 NOK
BELAF378790000
SP Bel-Art® sampling syringe
Prøvetakere Væskeprøvetakere
Sample liquids by simply squeezing the bulb, submersing in liquid, and releasing to draw liquids into the syringe.
  • Can be inverted to contain liquid in the graduated bulb
  • Graduation in 5 ml increments
  • Reusable item can be unscrewed from bulb for easy cleaning and storage
Sertifiseringer: FDA grade materials.
NB: Not recommended for use with liquids whose temperature is higher than 80 °C.
*Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904