Desiccators, vacuum, SP Bel-Art Techni-Dome®
Leverandør: Bel-Art Products, a Part of SP
F420290000
BELAF420290000EA 13575 NOK
BELAF420290000
Desiccators, vacuum, SP Bel-Art Techni-Dome®
Eksikatorer Eksikator, plast
This domed desiccator provides a spacious interior for large, moisture sensitive materials. Laboratory grade polycarbonate provides a clear, unobstructed view of contents.
  • Two removable PP shelves with Ø 365 and 225 mm
  • Side handles provide easy portability
  • Gas ports can be closed to allow use with standard desiccants
Interior volume of 65 L, holds items up to 450 mm tall or wide, including racks, trays and instruments. Tapered stopcock accepts ¼ to ³/₈" int.Ø and have easy turning PTFE valve to facilitate the turbulence-free entrance of gases,More Product Information