Desiccator plates, high heat, Minerit HD, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
F420380230
BELAF420380230EA 4500 NOK
BELAF420380230
Desiccator plates, high heat, Minerit HD, SP Bel-Art
Eksikatorer Eksikatorplater
Withstand temperatures up to 149 °C (300 °F). Contain no asbestos and are non combustible, stable and chemical resistant. Circular and perforated with 6 mm diameter holes on 25 mm centers in a straight line pattern.
Bestill nå
SPESIFIKASJONER
Tykkelse