Desiccators, vacuum, with plate, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
F420430000
BELAF420430000EA 4500 NOK
BELAF420430000
Desiccators, vacuum, with plate, SP Bel-Art
Eksikatorer Eksikator, plast
Ideal for vacuum drying, degassing and teaching demonstrations, this vacuum chamber can withstand a full vacuum at room temperature.
  • One piece O-ring seals the clear polycarbonate top to the white polycarbonate base plate
  • Polypropylene stopcock with Teflon® PTFE plug and tapered hose barb accepts tubing with 6 mm int.Ø.