Autoclave deodoriser, triple strength, Odo-Clave®, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
H131990000
BELAH131990000EA 5575 NOK
BELAH131990000
Autoclave deodoriser, triple strength, Odo-Clave®, SP Bel-Art
Deodoranter
This heat-released deodorant pads neutralise autoclave odors with a pleasant scent.
  • Simply open foil pouch and place pad in bag prior to autoclaving
  • Pads have a three year life span; will last longer if stored in a sealed container
  • Triple strength clove releases three times the deodorant of the standard pads while costing less than two standard pads
W×L: 57×57 mm
Forpakning: Packed 10 pads per bag, 10 bags per box.