Slide caliper, Digi-Max™, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
H134170000
BELAH134170000EA 11000 NOK
BELAH134170000
Slide caliper, Digi-Max™, SP Bel-Art
Skyvelær
Measurements taken with push button selection between metric and English scales and a self-zeroing feature for accuracy of ±0,1 mm.
  • LCD display
  • On/Off button preserves battery life
  • Metric scale measures from 0 to 150 mm and English scale from 0 to 6"
Length: 224 mm
Leveringsinformasjon: Supplied in a protective plastic case.