Levelling table, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
H183100000
BELAH183100000EA 5825 NOK
BELAH183100000
Levelling table, SP Bel-Art
Benkepapir Benkepapir
This clear acrylic level surface helps when pouring immunological plates, horizontal agarose gels and acrylamide sequencing gels.
  • Each leg is adjustable
  • Inset levelling bulb below the working surface assists in accurate adjustment