Pipette stand, Poxygrid®, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
H189620000
BELAH189620000EA 3800 NOK
BELAH189620000
Pipette stand, Poxygrid®, SP Bel-Art
Pipettestativ
This epoxy-coated aluminum rack stores pipettes and protects pipette tips.
  • Holds up to 10 round shape pipettors such as Finnpipette®, Socorex®, Eppendorf® and other popular brands; two end holes fit Pipette-Aid® pipette fillers
  • Two stepped rows of 5 each
  • Second lower support row has smaller holes to accept tapered ends of the pipettors to keep them vertical
  • Quick and easy access for more efficient work; no more searching, fumbling or breakage