Pipette stands, Poxygrid®, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
H189620001
BELAH189620001EA 3175 NOK
BELAH189620001
Pipette stands, Poxygrid®, SP Bel-Art
Pipettestativ
These pipette stands made of epoxy coated aluminium provide storage and easy access for up to 10 Gilson Pipetman® pipettors plus up to two Pipet-Aid® pipettes.
  • Keep pipettors organised, clearly visible and ready for use
  • Epoxy coating is corrosion resistant and easy to clean
  • Available with extended base with raised lip to hold plastic tip boxes
  • For wall mouting or benchtop use