Vacu-rack, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
H189770000
BELAH189770000EA 10350 NOK
BELAH189770000
Vacu-rack, SP Bel-Art
Stativer for rør Tube Racks
PP.
  • Accepts tubes up to 102 mm long
  • Coloured tube stopper suspends the tube in its channel and offers immediate visual color identification
  • Three side openings support needle holders
  • Chemical and corrosion resistant
This rack is an ideal accessory for blood drawing. Keep 1 or 2 channels open for stacking blood samples in sequential order. Organise 143 vacuum-type syringe tubes; 80 tubes 12,7 mm (¹/₂") Ø and 63 tubes 16 mm (⁵/₈") Ø.