Sampling spoons, double-ended, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
H367110000
BELAH367110000EA 1375 NOK
BELAH367110000
Sampling spoons, double-ended, SP Bel-Art
Skjeer
Staninless steel, Fluo-Kem® PTFE coating. This spoon/spatula combination can be used for sampling and scraping.
  • Corrosion resistant stainless steel construction is strong and durable
  • Fluo-Kem® PTFE/FEP coating is FDA compliant and prevents material from sticking
  • Spatula size: 8×51 mm
Ideal for use in compact workstations or with balances.