Sampling spoon, large, Sterileware®, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
H369480000
BELAH369480000EA 2350 NOK
BELAH369480000
Sampling spoon, large, Sterileware®, SP Bel-Art
Skjeer
PS, white. This large disposable spoon made from FDA-grade PS is ideal for sampling semi-solids, viscous liquids or bulk materials.
  • Long handle reaches into containers and keeps hand away from sample
  • Each one-piece spoon is sealed in a polyethylene bag, ready for use
  • Sterilised by Gamma Irradiation
  • Lot stamped for accurate lot tracing, and shipped with a Certificate of Processing for Sterilisation