SP Bel-Art® precisionware sampler for acids
Leverandør: Bel-Art Products, a Part of SP
H378010000
BELAH378010000EA 5475 NOK
BELAH378010000
SP Bel-Art® precisionware sampler for acids
Prøvetakere Væskeprøvetakere
PE bottle threads on to end of rod to collect samples. Safely distances user from dangerous chemicals.
  • Fluid enters through fitting above bottle
  • T-handle for secure grip when lowering sampler
  • Handle has two piece threaded joint to shorten during transport or storage
Designed for hydrochloric, sulfuric and acetic acids, formaldehyde potassium or sodium hydroxide, in tanks rail cars, and other large vessels.