Bellows samplers, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
H378230050
BELAH378230050EA 2125 NOK
BELAH378230050
Bellows samplers, SP Bel-Art
Pipetter Pasteurpipetter og overføringspipetter
These bellows samplers made of PE provide quick and easy liquid sampling with one hand operation.
  • FDA grade polyethylene sampler tube
  • Long tapered tip with a 4 mm opening
  • Easy one-hand operation
Forpakning: Five per bag