SP Bel-Art® liquid sampler
Leverandør: Bel-Art Products, a Part of SP
H378270000
BELAH378270000EA 9400 NOK
BELAH378270000
SP Bel-Art® liquid sampler
Prøvetakere Væskeprøvetakere
Liquid sampler made of nickel-plated brass cup and tubular handle with finger grip. Cup is not replaceable and can not detach.
Suitable for immersion into a can, bottle, tank, vat ...