SP Bel-Art® three zone owder samplers
Leverandør: Bel-Art Products, a Part of SP
H378290000
BELAH378290000EA 7500 NOK
BELAH378290000
SP Bel-Art® three zone owder samplers
Prøvetakere Prøvetakere til faststoff
Three zone powder samplers.
  • Each of the three cells gathers samples
  • Fine point to sample the bag contents
  • Minimum damages to the bag
Used in non-homogenous powders, pellets, and other friable materials.