Pasteur pipettes, disposable
Leverandør: BELLCO GLASS
1277-10009
BELC1277-10009EA 4200 NOK
BELC1277-10009
Pasteur pipettes, disposable
Pipetter Pasteurpipetter og overføringspipetter
Made of borosilicate glass.
Suitable for the transfer of all infectious materials, culture media and for routine sampling.