Microscope slides with printed rings
Leverandør: BELLCO GLASS
5638-01920
BELC5638-01920EA 5550 NOK
BELC5638-01920
Microscope slides with printed rings
Objektglass Objektglass til mikroskop
These microscopy slides with three printed rings allow for counting bacteria by spreading a known volume of solution over a known area for the calculation of bacteria per sample.
Area (ring): 1 cm²