Polarimeter tubes and spare parts
Leverandør: BELLINGHAM STANLEY
35-58 35-57 35-46 35-47 35-68 35-60 35-64 35-29 35-30 35-11 35-20 35-21 35-91 35-95 35-96 35-90 37-540 37-541 35-17 35-18 37-543
635-0029EA 4625 NOK
635-0029 635-0028 635-0026 635-0027 635-0030 635-0289 635-0290 635-0024 635-0025 BELL35-11 635-1004 635-1005 BELL35-91 BELL35-95 BELL35-96 BELL35-90 BELL37-540 BELL37-541 BELL35-17 BELL35-18 BELL37-543
Polarimeter tubes and spare parts
Polarimetere
Polarimeter tubes are manufactured to high quality standards where tube ends are precision ground and windows made from selected low strain glass providing highest accuracy along the light path.
  • Jacketed funnel flow-through tubes for sugar testing laboratories
  • Water-jacketed tubes can be used to provide temperature control as required
  • Continuous flow through tubes allows monitoring of pumped samples
Sertifiseringer: Conforming to ICUMSA recommendations.
Bestillingsinformasjon: XPC adaptors and cover glasses for UV measurements are also available.