Digital polarimeters, ADP600
Leverandør: BELLINGHAM STANLEY
37-61 37-62 37-63 37-64 37-65 37-66
BELL37-61EA 0 NOK
BELL37-61 BELL37-62 BELL37-63 BELL37-64 BELL37-65 BELL37-66
Digital polarimeters, ADP600
Polarimetere
The ADP600 high precision polarimeters with internal Peltier temperature control are capable of measuring optical rotation to four decimal places angular across the UV/Visible spectrum using standard glass or low volume leur taper flow-through tubes.
  • Touch screen facilitates navigation through user and supervisor menus
  • Standard and low volume sample tubes
  • Single, double, or multiple wavelengths for UV/Visible applications
  • GLP software including date/time/batch
Scientists wishing to measure optical rotation, specific rotation or concentration ofMore Product Information